Print…
Polypropylene (PP) High-Flow Chemical-Resistance Centrifugal Pumps
Fornecedor: Avantor Fluid Handling
72013-05 72013-20 72013-00 72013-10
MFLX72013-05EA 546 EUR
MFLX72013-05 MFLX72013-20 MFLX72013-00 MFLX72013-10
Polypropylene (PP) High-Flow Chemical-Resistance Centrifugal Pumps
Bombas Bombas centrífugas
Twin bearing system handles thrust loads for extended service life.
  • Quiet, energy efficient, and compact
  • Internal cooling circuit reduces bearing temperature
  • Enclosed motor for use in harsh environments
  • All wetted parts are nonmetallic for excellent chemical resistance
High-flow polypropylene pumps utilize an open impeller design to produce high flows with very little horsepower.
Fornecimento: Includes a 6-ft (1.8 m) cord with stripped ends.