Print…
Masterflex® Adapter Fittings, Luer to Luer, Straight, Avantor®
Fornecedor: Avantor Fluid Handling
45508-22
MFLX30800-14EA 39.3 EUR
MFLX30800-14 MFLX45508-80 MFLX45508-82 MFLX45508-22
Masterflex® Adapter Fittings, Luer to Luer, Straight, Avantor®
Conexões e Conectores
Full material traceability to ensure process integrity.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include