Print…
Masterflex® Single-Use Flow Sensor Outputs, Avantor®
Fornecedor: Avantor Fluid Handling
MFLX24507-17EA 423 EUR
MFLX24507-17 MFLX24507-18 MFLX24507-25 MFLX24507-26
Masterflex® Single-Use Flow Sensor Outputs, Avantor®
Medidores de fluxo
Build a complete single-use flow sensor system customizable to your application.
  • Modular interchangeable flow sensor systems with single-use design
  • Quickly connect 5 to 30 V square wave pulse output for easy measurement readings
The Masterflex® single-use flow sensor system is comprised of four components. Select the flow sensor, flow tubes, barcode scanner, and output that suit your application needs for a customizable approach to single-use flow measurement.
Four output options are available — digital to analog converter, metering pump control system,Mais informação do produto