Print…
Masterflex® L/S® Digital Process Drives, Avantor®
Fornecedor: Avantor Fluid Handling
MFLX07575-30-EUEA 7010 EUR
MFLX07575-30-EU MFLX07575-40-EU
Masterflex® L/S® Digital Process Drives, Avantor®
Bombas Acionamentos de bombas Acionamentos de bombas peristálticas
Transfer, dispense, and meter in lab, process and washdown environments.
  • Flow rate 0.006 to 3400 ml/min with L/S tubing—flow rate depends on drive rpm pump head, and tubing size
  • Digital interface with graphical LCD shows pump performance and operating mode— continuous run, timed dispense, copy dispense, or volume dispense
  • Remote control available using analog communication—voltage, current and contact closure
  • Dispense by Volume in ml (0.001 to 9,999 mL), L, or gal; Copy—1 to 9,999 dispense cycles; or Time—time interval of on/off cycles (1 sec to 99 hr, 59
Mais informação do produto