Print…
Masterflex® L/S® Multichannel Pump Heads, Avantor®
Fornecedor: Avantor Fluid Handling
MFLX07534-04EA 1540 EUR
MFLX07534-04 MFLX07534-08 MFLX07535-04 MFLX07535-08 MFLX07536-02 MFLX07536-04
Masterflex® L/S® Multichannel Pump Heads, Avantor®
Bombas Cabeças de bomba Cabeças de bomba peristáltica
Get low-pulsation synchronous flow in multiple channels without cartridges.
  • Flow range 0.045 to 2300 ml/min per channel— flow rate depends on drive rpm and tubing size
  • Stainless steel (SS) rotor / roller for greater durability and chemical resistance
  • Stackable two-, four-, or eight-channel heads for up to 24 channels
  • Simple tubing installation—lLoad the tubing, latch the occlusion bed, and run the pump
The L/S® multichannel pump heads are designed with machined stainless steel (SS) and anodized aluminum with SS rotor, bearings, and rollers forMais informação do produto