Print…
Masterflex® L/S® Fixed-Speed VDC Compact Drives, Avantor®
Fornecedor: Avantor Fluid Handling
MFLX07533-50EA 1040 EUR
MFLX07533-50 MFLX07533-60 MFLX07533-70 MFLX07533-80
Masterflex® L/S® Fixed-Speed VDC Compact Drives, Avantor®
Bombas Acionamentos de bombas Acionamentos de bombas peristálticas
Take it where you need it—ideal for field use.
  • Flow rate 6 to 2700 ml/min with L/S tubing—flow rate depends on drive rpm, pump head and tubing size
  • Rugged, continuous-duty motor is ideal for applications that require full-time duty cycle
  • Vary input voltage to change drive speed and reverse input voltage polarity to reverse direction—Control drive completely using power source
  • Constructed with chemical-resistant epoxy-coated steel motor housing for IP52 rating
These fixed-speed compact drives operate on 12 VDC input power. Wire power from your 12 or 2Mais informação do produto