Print…
Masterflex® L/S® Precision Process Drives, Avantor®
Fornecedor: Avantor Fluid Handling
MFLX07528-40-EUEA 4480 EUR
MFLX07528-40-EU MFLX07528-50-EU
Masterflex® L/S® Precision Process Drives, Avantor®
Bombas Acionamentos de bombas Acionamentos de bombas peristálticas
Easy-to-use drives offer precise speed control and digital display of rpm—NEMA 4X housings stand up to process and washdown applications.
  • Flow rate 0.06 to 3400 ml/min (depends on drive rpm and tubing size)
  • Four digit LED display shows motor rpm—confirm speed setting at a glance
  • Integrated open-head sensor stops drive motor when pump head is opened
  • Reversible motor—purge tubing before or after pumping; pump fluid in either direction
  • LED indicators confirm motor direction
  • Powerful motors let you stack 2 or 4 pump heads for increased flow rates and for
Mais informação do produto