Print…
FMI Series RH Valveless Piston Pump Heads
Fornecedor: Avantor Fluid Handling
07117-61 07117-64
MFLX07117-53EA 1270 EUR
MFLX07117-53 MFLX07117-55 MFLX07117-63 MFLX07117-65 MFLX07117-69 MFLX07117-71 MFLX07117-61 MFLX07117-64
FMI Series RH Valveless Piston Pump Heads
Bombas Cabeças de bomba Cabeças de bomba de pistão
Small displacement provides excellent precision.
  • Fully adjustable from zero to maximum
Valveless piston pumps are low maintenance and suffer fewer piston siezures or clogs. With a single moving part, there is less drift over time, offering millions of cycles of reliable, repeatable pumping.