Print…
Stopcocks with Flow Indicator, PVC
Fornecedor: Avantor Fluid Handling
06225-52 06225-54 06225-56 06225-50
MFLX06225-52EA 46.8 EUR
MFLX06225-52 MFLX06225-54 MFLX06225-56 MFLX06225-50
Stopcocks with Flow Indicator, PVC
Torneiras
Easily control flow with flow indicator handle.
  • Constructed of durable PVC and EPDM
PVC bodies, EPDM seals and seats. Flow indicator on handle. Maximum pressure is 150 psi at the maximum temperature of 120 °F (49 °C); maximum vacuum is 29.9" Hg.