Print…
Paraffin ovens with natural convection, universal, UNpa series
Fornecedor: MEMMERT
UN75PA UN110PA UN160PA UN30PA UN55PA
MEMMUN75PAEA 3510 EUR
MEMMUN75PA MEMMUN110PA MEMMUN160PA MEMMUN30PA MEMMUN55PA
Paraffin ovens with natural convection, universal, UNpa series
Estufas Estufas standard
UNpa paraffin ovens provide precise and homogenous temperature control of paraffin embedding medium without any loss in quality. UNpa paraffin ovens are designed specifically for reliable sample preparation in the laboratory and have an intuitive and easy to use operating menu plus a high-quality, corrosion-resistant and easily cleanable stainless steel working chamber and housing.
  • High degree of thermal safety through multiple overtemperature protection
  • Intuitive, user friendly ovens with 2 TFT displays. ControlCOCKPIT parameters: Temperature (Celsius or
Mais informação do produto