Print…
Sterilisers, SF models
Fornecedor: MEMMERT
SF110 SF160 SF450 SF260 SF30 SF55 SF75 SF750
MEMMSF110EA 2460 EUR
MEMMSF110 MEMMSF160 MEMMSF450 MEMMSF260 MEMMSF30 MEMMSF55 MEMMSF75 MEMMSF750
Sterilisers, SF models
Esterilizadores Esterilizadores por ar quente
This steriliser complies with all relevant standards and regulations for medical products and guarantees reliable programme sequences, irrespective of the load and chamber volume. Especially due to the SetpointWAIT function the validation of this hot air steriliser is possible anytime. The SetpointWAIT function ensures that the programme is only continued when the setpoint temperature has been reached. Precisely maintaining the sterilisation time and thus completely killing off even highly resistant microorganisms is therefore guaranteed at all times, irrespective of the sterilisation loadMais informação do produto