Print…
Urine collection tubes, primary, with screw cap, WHEATON®
Fornecedor: DWK Life Sciences
327220 327230
LSUK327220EA 195 EUR
LSUK327220 LSUK327230
Urine collection tubes, primary, with screw cap, WHEATON®
Sistemas para diagnóstico Clinical Sample Collection/Preparation Coleta/Preparação de Amostras de Urina
These WHEATON® round bottom tubes are manufactured from virgin PS. The tubes have a capacity of 11 ml and are fitted with a leakproof screw cap for ease of transportation and storage of the sample. The tubes are supplied with a collection cup for the patient’s comfort during sample collection, and for them to easily transfer the sample to the tube.
  • Ideal for use with automated urine analysis machines
  • Printed label for patient details
  • Option of plain tube or pre-dosed with boric acid
  • Removes the need for unnecessary decanting of sample in the laboratory
Mais informação do produto