Print…
Culture Media in Bottles
Fornecedor: LIOFILCHEM
495270
LIOF495270EA 154 EUR
LIOF495270
Culture Media in Bottles
Meios de cultura Meios Preparados
Buffered NaCl Peptone Solution pH 7.0 is used for dissolving, suspending and diluting test samples.
  • Its composition conforms to recommendations in the Harmonized Pharmacopoeias for the microbiological examination of non sterile products
  • This solution also conforms to ISO 21149 for the detection of viable microorganisms in cosmetics
  • Shelf life two years
The powder is very hygroscopic, store the powder at 10 to 30 °C, in a dry environment, in its original container tightly closed. Store bottles and bags at 10 to 25 °C away from light. Do not use the product beyond its expiry date on the label or if product shows any evidence of contamination or any sign of deterioration.