Print…
Pharmaceutical Perfection refrigerators compliant with DIN 13277, HMFvh
Fornecedor: Liebherr
HMFVH4001 HMFVH4011 HMFVH5501 HMFVH5511
LIEBHMFVH4001EA 3303.9 EUR
LIEBHMFVH4001 LIEBHMFVH4011 LIEBHMFVH5501 LIEBHMFVH5511
Pharmaceutical Perfection refrigerators compliant with DIN 13277, HMFvh
Frigoríficos
Ultra-efficient pharmaceutical refrigerators have been designed for the particular requirements with their dynamic cooling system, highly effective insulation, and precise temperature control, they guarantee reliable storage of sensitive substances and preparations.
  • Integrated data logger and USB interface for reading the data logger
  • PT1000 product sensor and Sensor feedthrough
  • Min./max. temperature log shows whether the storage temperatures were within the tolerance limits
  • Insulated and glass door models with vertical lighting, self-closing door at <90°