Print…
Flat Bags
Fornecedor: KWP KUNSTSTOFF WERK PLUR
123045
KWPA123045EA 0 EUR
KWPA123045
Flat Bags
Sacos Sacos para Armazenamento
Flat bag made from LDPE.
  • Suitable for cleanroom applications (ISO class 5)
  • LDPE grade for pharmaceutical applications