Print…
Software, EuroOsmo® 7400
Fornecedor: KNAUER
A3705
KNAUA3705EA 1030 EUR
KNAUA3705
Software, EuroOsmo® 7400
Software Programas de Laboratório
EuroOsmo® 7400 is a user friendly software developed for the freezing point osmometers K-7400 and K-7400S.
  • Data can optionally be exported into xls or txt files
It allows control of the instruments and recording of the measured data. The software automatically plots the temperature curve for each measurement and calibration and allows saving of the measured values. As with the instrument itself, the user can choose between freezing point depression or osmolality to be displayed also in the software. The data can optionally be exported into xls or txtMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable