Print…
Disposable 12 mm generator, Polytron® PT-DA 12/2SYN-E123
Fornecedor: KINEMATICA
11030035
KINE11030035EA 934 EUR
KINE11030035
Disposable 12 mm generator, Polytron® PT-DA 12/2SYN-E123
Homogeneizadores Hastes de Dispersão
Disposable/reusable generators for when processing multiple samples per day, where the required sterilisation of stainless steel generators represents a significant time limitation.
  • Prevention of cross-contamination
  • Standard dispersing applications