Print…
Test tubes for pediatric blood takings with collection rim
Fornecedor: KIMA SAS DI CHIARIN RENZO
812550 812020 813010 813505 813805 814074 813510 810176 813005 813030 811020 810020
720-2759EA 26.7 EUR
720-2759 720-2758 720-2699 720-2700 720-2702 720-2703 720-2701 KIMA810176 KIMA813005 KIMA813030 KIMA811020 KIMA810020
Test tubes for pediatric blood takings with collection rim
Sistemas para diagnóstico Clinical Sample Collection/Preparation Coleta/Preparação de Amostras de Sangue
PP, with transparent label
Microtest sterile pediatric test tubes size 14×45 mm. These test tubes can be adapted to an extender to bring the overall dimensions to 13×75 mm. This device enables use in standard centrifuges and preanalytical equipments.