Print…
Frascos de fundo redondo, com juntas com esmerilado padrão
Fornecedor: KARTELL
1540
KART1540EA 263 EUR
KART1540
Frascos de fundo redondo, com juntas com esmerilado padrão
Balões Balões de Fundo Redondo
PFA, translucent.
  • Very good chemical resistance
  • Excellent thermal resistance from −200 to +250 °C
  • Vacuum: suitable
For rotary evaporators and distillation of fine chemicals.