Print…
M9400 Wraplite Safety spectacle
Fornecedor: JSP
ASA718-161-100 ASA718-1B1-124
JSPAASA718-161-100EA 5.3 EUR
JSPAASA718-161-100 JSPAASA718-1B1-124
M9400 Wraplite Safety spectacle
Óculos
Single lens, wraparound, comfortable and lightweight lens design provides excellent field of vision. A popular choice of spectacle that can be worn for long periods of time with continual user comfort ensured.
  • Lightweight
  • Wraparound design
  • Blocks 99,9% of ultraviolet radiation
  • Anti-scratch and anti-mist models available
Single lens wraparound safety spectacles offer protection against low energy, high-speed particles at 45 m/s (101 mph). Anti-scratch and anti-mist treatments are available to provide increased lens efficiency.
Certificações: Mais informação do produto