Print…
Protease inhibitor cocktails I, animal-free
Fornecedor: Thermo Fisher Scientific
J65358.LQ
J65358.LQEA 66.5 EUR
J65358.LQ
Protease inhibitor cocktails I, animal-free
Cultura celular, Suplementos e aditivos
A 1X stock solution contains 500 μM AEBSF.HCl, 150 nM aprotinin, 1 μM E-64, 0,5 mM EDTA and 1 μM leupeptin hemisulfate. Reconstitute each vial with 1 ml water to obtain a 100X stock solution.