Print…

Cilostamide

Fornecedor: Thermo Fisher Scientific

Sinônimos: N-Cyclohexyl-N-methyl-4-((2-oxo-1,2-dihydroquinolin-6-yl)oxy)butanamide

J65319.MAEA 136 EUR
J65319.MA
Cilostamide
Cilostamide

CAS No.: 68550-75-4 Molecular Formula: C20H26N2O3 Formula Weight: 342.43 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White MDL No.: MFCD00673958


Applications: A cell-permeable selective inhibitor of cGMP-inhibited phosphodiesterase

Fórmula: C₂₀H₂₆N₂O₃
Peso Molecular: 342.43 g/mol
Número MDL : MFCD00673958
Número CAS: 68550-75-4

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR