Print…
Reagente de Schiff
Fornecedor: Thermo Fisher Scientific
Atenção
J62171.AP
J62171.APEA 126 EUR
J62171.AP
Reagente de Schiff
Schiff reagente
Used for detection of glycoproteins in polyacrylamide gels.
Intended for use in Sigma Periodic Acid Staining Procedure, 395. Kit is intended for use with blood, bone marrow, tissue touch preparations or routine tissue sections. It is used for staining glycoproteins in polyacrylamide gels. Schiff's reagent is also sometimes used to dye organic tissues. It is been widely used for detection of a certain functional group in chemical solutions.
Used for detection of glycoproteins in polyacrylamide gels