Print…
High technology colony counter, Scan® 500
Fornecedor: INTERSCIENCE
436000
INSC436000EA 0 EUR
INSC436000
High technology colony counter, Scan® 500
Contadores de colónias
High technology colony counter with software that can be linked to a PC via a FireWire connection.
  • Minimum size of colonies is 0,1 mm
  • Detects up to seven colours on the same Petri dish
  • Measurement of the inhibition zone
Can count colonies on a Petri dish in less than one second for a fast, accurate, and traceable reading of the result.
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Tamanho da caixa Petri dishes (55 – 90 mm) and Spiral® plating
Alimentação 240 V/50 – 60 Hz
Ampliação
Câmara CMOS colour camera
Resolução 640×480 pixels
W×D×H 280×270×290 mm
Peso 8,4 kg