Print…
Remote Control, WiCo T 25 easy clean
Fornecedor: IKA
25006310
IKAA25006310EA 1025 EUR
IKAA25006310
Remote Control, WiCo T 25 easy clean
Homogeneizadores Homogeneizadores de Bancada
The WiCo wireless remote control allows user to operate and monitor the disperser T 25 easy clean control e.g. in a fume hood and therefore helps protect the user from exposure to toxic materials.
  • WiCo is only compatible with T 25 easy clean control disperser
Fornecimento: Supplied with wall mount WH 1 1 as well as two USB cables.