Print…
Immersion Circulators, ICC 100 lite eco 8
Fornecedor: IKA
10013176
IKAA10013176EA 1530 EUR
IKAA10013176
Immersion Circulators, ICC 100 lite eco 8
Termóstatos de circulação Sistemas de circulação aberta Controladores do termostato
The compact immersion circulators of the lite series are powerful and of high quality, with a focus on essential basic functions, excellent performance and efficient operation. ICC 100 lite is suitable for tempering liquids up to 100 °C and internal circulation.
  • Max. temperature in the plastic bath (eco): 100 °C (water only)
To make configuration of your temperature control system as convenient as possible, IKA offers 'ready-to-connect' packages with all the necessary accessories.
Fornecimento: Package includes: ICC 100 lite compact immersionMais informação do produto