Print…
Transformation kit, EZ-Yeast™, MP Biomedicals
Fornecedor: MP Biomedicals
112100200
ICNA112100200EA 108 EUR
ICNA112100200
Transformation kit, EZ-Yeast™, MP Biomedicals
Reagentes de ácidos nucleicos Reagentes de Transformação
The EZ-Yeast™ Transformation Kit is designed for high throughput transformation with efficiencies up to 10³ transformants per μg plasmid DNA. It is ideal for cases where a large number of transformations must be performed, but only a few transformants are needed. Perfect for a quick analysis of potentially positive two-hybrid colonies, it does not require fully competent cells prior to transformation.
Fornecimento: Kit contains EZ-Transformation Solution and carrier DNA.