Print…

2'-Deoxycytidine hydrochloride, pó branco

Fornecedor: MP Biomedicals

Sinônimos: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one hydrochloride

0210148301 0210148305 0210148310
ICNA0210148301EA 56.9 EUR
ICNA0210148301 ICNA0210148305 ICNA0210148310
2'-Deoxycytidine hydrochloride, pó branco
2'-Deoxycytidine hydrochloride

Deoxycytidine is a deoxyribonucleoside, a component of deoxyribonucleic acid. It is similar to the ribonucleoside cytidine, but with one hydroxyl group removed from the 2' position.


  • Cell culture


Soluble in water (50mg/ml-clear colorless solution).


Atenção: Research Use Only (RUO).

Fórmula: C₉H₁₄ClN₃O₄
Peso Molecular: 263.68 g/mol
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00012840
Número CAS: 3992-42-5

Encomendar

Especificações dos Resultados do Teste

Identity Test Passes
FTIR Conforms to Standard


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR