Print…
Pressure dispense connector, Burdick & Jackson®, NP515
Fornecedor: Honeywell Chemicals
NP515
HONCNP515EA 1842.27 EUR
HONCNP515
Pressure dispense connector, Burdick & Jackson®, NP515
Dispensadores para Frascos
This connector allows for general purpose dispensing of solvent from B&J NOWPak II containers using up to 15 psig pressure.
It consists of:
- A cap connector with a 4" PFA elbow solvent fitting, a 4" inert gas elbow pressure fitting and a 15 psig pressure relief valve.
- An FEP diptube with a 40-micron stainless steel frit to prevent drips while changing containers.
- A 10' harness of 4" FEP solvent tubing and 4" nylon pressure tubing.
Pressure dispense connectors can easily attach stainless steel B&J NOWPak II containers to DNA synthesizers, peptide synthesizers and otherMais informação do produto