Print…
Strontium AAS Standards, (CRMs), Fluka™
Fornecedor: Honeywell Chemicals
E4072-100ML E4072-500ML E4073-100ML E4073-500ML E4401-100ML
HONCE4072-100MLEA 38.04 EUR
HONCE4072-100ML HONCE4072-500ML HONCE4073-100ML HONCE4073-500ML HONCE4401-100ML
Strontium AAS Standards, (CRMs), Fluka™
Padrões Cationic Standards Strontium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificações: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).