Print…
Molybdenum AAS standards, (CRMs), Fluka™
Fornecedor: Honeywell Chemicals
E4046-100ML E4046-500ML E4047-100ML E4047-500ML
HONCE4046-100MLEA 34.01 EUR
HONCE4046-100ML HONCE4046-500ML HONCE4047-100ML HONCE4047-500ML
Molybdenum AAS standards, (CRMs), Fluka™
Padrões Cationic Standards Molybdenum Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certificações: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).