Print…

Dietiléter, dried (max. 0.0075% H₂O) Stabilised, Puriss. p.a. ACS, Riedel-de Haën™

Fornecedor: Honeywell Chemicals

Sinônimos: Eter etílico

34942-18L
HONC34942-18LEA 2115.32 EUR
HONC34942-18L
Dietiléter, dried (max. 0.0075% H₂O) Stabilised, Puriss. p.a. ACS, Riedel-de Haën™
Dietiléter
Fórmula: (CH₃CH₂)₂O
Peso Molecular: 74.12 g/mol
Ponto de ebulição: 34…35 °C (1,013 hPa)
Ponto de Fusão: –116 °C.(1,013 hPa)
Densidade: 0.71 g/cm3 (20 °C)
Ponto de Inflamação: –45 °C (1,013 hPa.)
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00011646
Número CAS: 60-29-7
UN: 1155
ADR: 3 I
Merck Index: 12,03852

Encomendar

Especificações dos Resultados do Teste

ExplosionLimit 36.5 %
Assay (GC) Min. 99.5 %
Boiling range 34 - 35 °C
Density (D 20/4) 0.713 - 0.714
Non-volatile matter Max. 0.001 %
Water (Karl Fischer) Max. 0.0075 %
Free acid (as CH₃COOH) Max. 0.0002 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.000005 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000005 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.000005 %
Acetone (GC) Max. 0.005 %
Aldehydes complying
Carbonyl compounds (as CO) Max. 0.001 %
Ethanol (GC) Max. 0.02 %
Methanol (GC) Max. 0.02 %
Peroxides (as H₂O₂) Max. 0.00003 %
Reaction against H₂SO₄ complying
APHA Max. 10


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR