Print…
Avantor® Hichrom HI-BIODIESEL 103, GC Columns
Fornecedor: Avantor
HI39-32-025-30EA 1050 EUR
HI39-32-025-30
Avantor® Hichrom HI-BIODIESEL 103, GC Columns
Cromatografia, Colunas
Application specifc column for free and total glycerine (phase stable up to 380 °C) and for FAMEs in biodiesel analysis.
Max temperature limits may change depending on the film thickness.
Encomendar
ESPECIFICAÇÕES
Phase name HI-BIODIESEL 103
Environmentally Preferable
Phase composition Proprietary Phase developed for Biodiesel EN14103 analysis
Crossbond Yes
Temperature limit 380 °C