Print…
Blood Bank Refrigerators, TSB
Fornecedor: Thermo Scientific
TSB140REGBV TSB250REGBV TSB700REGBV TSB1500REGBV
HERATSB140REGBVEA 6070 EUR
HERATSB140REGBV HERATSB250REGBV HERATSB700REGBV HERATSB1500REGBV
Blood Bank Refrigerators, TSB
Frigoríficos
The Thermo Scientific TSB series blood bank refrigerators are designed to meet the strict requirements for storage of blood and blood related products in full compliance with 93/42/CE - class IIa regulation.
  • Touchscreen digital controller with onboard datalogging
  • Whisper quite operation <50 dBA
  • Triple pane glass doors with steel cabinet material
  • White painted interior and exterior
  • Temperature, door ajar, and power failure (with battery backup) alarms
  • Low energy consumption with real time energy monitoring
  • Low heat output
  • Electronic lock with password
Mais informação do produto