Print…
Ultra low temperature freezers, with 4 inner doors, –86 °C, TDE series
Fornecedor: Thermo Scientific
TDE30086FVULTS TDE40086FVULTS TDE50086FVULTS TDE60086FVULTS
HERATDE30086FVULTSEA 12100 EUR
HERATDE30086FVULTS HERATDE40086FVULTS HERATDE50086FVULTS HERATDE60086FVULTS
Ultra low temperature freezers, with 4 inner doors, –86 °C, TDE series
Congeladores Congeladores de temperaturas ultra baixas
Thermo Scientific™ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models, maximizing storage capacity from 30000 up to 60000 2 ml vials. The TDE Series is a sustainable and reliable solution with ultimate sample security and operational savings for every laboratory. Sustainability features include natural refrigerants and water-blown foam, compliant with many sustainability standards.
  • Powered by H-drive, save up to 25% in energy usage and heat rejection over conventional refrigerant ultra-low freezers
  • Easy-to-use set-up