Print…
Hei-TORQUE Overhead Stirrers, Gold 1 Package
Fornecedor: Heidolph Instruments GmbH & Co.KG
501-61029-00
HEID501-61029-00EA 2570 EUR
HEID501-61029-00
Hei-TORQUE Overhead Stirrers, Gold 1 Package
Agitadores magnéticos e suspensos Agitadores suspensos
In order to provide the perfect end-to-end solution for powerful stirring and the simplest operation in the laboratory, we have added packages to the Hei-TORQUE product group.Each package contains a telescopic stand and a suitable clamp for easy use on lab tables.
Fornecimento: Hei-TORQUE Ultimate 100 comes with telescope stand and clamp.