Print…
Assistent Brush for Beakers
Fornecedor: GLASWARENFABRIK KARL HECHT
41936010
HECH41936010EA 282 EUR
HECH41936010
Assistent Brush for Beakers
Escovilhões
Wood handle, natural bristle brush designed for use with beakers, or similar sized laboratory glassware.
  • Wood handle
  • Natural bristles
Natural bristles are ideal for cleaning beakers, glass or plastic cups, measuring cups and more.