Print…
Distilling flask with side tube, Assistent
Fornecedor: GLASWARENFABRIK KARL HECHT
41037016
HECH41037016EA 22.2 EUR
HECH41037016
Distilling flask with side tube, Assistent
Balões Frascos de Destilação
This distilling flask is made of borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion
The length of the side-tube is approximately 200 mm.