Print…
Cooling boxes, True North®
Fornecedor: HEATHROW
HS28714K HS28724B HS28721G HS28729R
HEATHS28714KEA 111 EUR
HEATHS28714K HEATHS28724B HEATHS28721G HEATHS28729R
Cooling boxes, True North®
Caixas Refrigeradores de bancada
Cooling containers with nest lid beneath the bucket or pan are made of rigid single-molded polyurethane to provide the durability needed for chilling temperature sensitive samples.
  • Precise pour spout and generous handles
  • Taper design for easy stacking and insulation properties for ultra cold
  • Close-fit lid keeps evaporation to a minimum and features a comfortable recessed knob
  • Lid and bottom of the bucket/pan are keyed to fit together
These boxes are ideal for ice, ice-salt slurries, and more. For low temperature use to −80 °C.