Print…
Freezer boxes (Cryoboxes), True North®
Fornecedor: HEATHROW
HS120371 HS120372 HS120373 HS120376 HS120388 HS120392 HS120367
HEATHS120371EA 84 EUR
HEATHS120371 HEATHS120372 HEATHS120373 HEATHS120376 HEATHS120388 HEATHS120392 HEATHS120367
Freezer boxes (Cryoboxes), True North®
Caixas Caixas de criotubos
True North® freezer boxes (cryoboxes) were designed with sustainability in mind and are made from a specially formulated corrugated or thin film polypropylene.
  • Easy self-assemble boxes shipped flat-packed
  • More durable than card boxes as they are not susceptible to moisture
  • Dividers included with the boxes
  • Mold resistant
  • Store down to temperatures of –80 °C (Corrugated PP boxes)/–196 °C (Thin PP film boxes)
Boxes are moisture resistant, which significantly reduces the chance of mold growth. With superior durability, polypropylene outlasts the cardboardMais informação do produto