Print…
Poly-spoons
Fornecedor: HAMMACHER KARL
HSN175-30 HSN176-40 HSN173-21 HSN174-25
HAMMHSN175-30EA 26.8 EUR
HAMMHSN175-30 HAMMHSN176-40 HAMMHSN173-21 HAMMHSN174-25
Poly-spoons
Colheres
Poly-spoons made of stainless-steel.
  • Double-ended: One spoon and one spatula end