Print…
Built-in syringe guide
Fornecedor: HAMILTON BONADUZ
86207
HAMI86207EA 226 EUR
HAMI86207
Built-in syringe guide
Seringas Seringas para cromatografia
The syringe guide comes with a syringe installed from the factory. This eliminates the assembly step which can lead to damage of the smaller volume syringes. This is especially critical for 7000 series syringes which have extremely small plunger wires.