Print…
Refrigerated circulating bath, VersaCool™
Fornecedor: THERMO ELECTRON LED
231131300
HAKA231131300EA 12100 EUR
HAKA231131300
Refrigerated circulating bath, VersaCool™
Termóstatos de circulação Sistemas de circulação fechada Circuladores refrigerados
The VersaCool™ Refrigerated circulating bath features a new headless design, suitable for use as a recirculating refrigerated bath or heated circulator to circulate fluid from the bath to specific applications.
  • More space to fit samples
  • Automatically detects appropriate voltage and works on all voltages worldwide
  • Lower power consumption and energy costs
  • Tool-less, drip-less rack
  • Left-handed hinge, right-handed hinge, or unhinged lid
  • Intuitive, 5,7" touchscreen colour display for easier and more efficient setup
  • Two year warranty
  • Ethernet RS232