Print…
ORP combination electrodes, IntelliCAL™ MTC101 / MTC301
Fornecedor: Hach
MTC10103 MTC10105 MTC10101 MTC10110 MTC10115 MTC10130 MTC30101 MTC30103
HACHMTC10103EA 433 EUR
HACHMTC10103 HACHMTC10105 HACHMTC10101 HACHMTC10110 HACHMTC10115 HACHMTC10130 HACHMTC30101 HACHMTC30103
ORP combination electrodes, IntelliCAL™ MTC101 / MTC301
Eléctrodos Redox Electrodes
Digital combination ORP/Redox probes with built-in temperature sensor. Ideal for measuring absolute mV values in wastewater, drinking water and general applications.
  • Compatible with digital Hach meters only (e.g. HQd meters)
  • Traceability in measurement history: Stored time and date stamp for each measurement, operator and sample ID, calibration history, parameter, and probe serial number
  • User alerted when re-calibration is needed
  • Plug & Play: Can be moved between meters without the need to re-calibrate or re-enter measurement settings
  • Rugged digital
Mais informação do produto