Print…
Sampler dippers
Fornecedor: Hach
2929302
HACH2929302EA 90.1 EUR
HACH2929302
Sampler dippers
Amostradores Colectores de amostra
Sampler dipper with PP cups for sampling from tanks, vats, streams, ponds, lakes.
  • Two pouring spouts
  • Cups with threaded fittings that screw onto the handle
  • Cups are autoclavable at 121 °C
The ends of the dipper are sealed for easy cleaning and include a hook for hanging.