Print…
Ionic strength adjustor for ammonia
Fornecedor: Hach
2824349
HACH2824349EA 80.1 EUR
HACH2824349
Ionic strength adjustor for ammonia
Electrólitos Eléctrodos, soluções enchimento
Ammonia Ionic Strength Adjustor (ISA) buffer solution for ammonia determinations by electrode methods.