Print…
Reagents for DR800, DR900, DR2010, DR2500, DR2400, DR2800, DR3800, DR3900, DR5000, DR6000
Fornecedor: Hach
1206499 1417099 2122332 2212129 2212242 2297255 2319900 2244700 2458200 2459200 2293666 2070042 2070142 2070242 2446600 2241732 2241832 2229726 2257169 2658300 2257700 2603100 2603101 2277800 2352449 2352512 2368531 2277924 2278024 2401066 2401268 2544800 2244600 2604299 2319800 2563000 2459000 2242300 2296600 2449400 199526 2651600 2658448 2293566 2770900 2790800 44449 2076053 2678500 2678600 2076049 2754048 2947553 HPT310 2368749 2368964 2368655 2368855 2105728 2527025 2658825
HACH1206499EA 125 EUR
HACH1206499 705-0558 HACH2122332 HACH2212129 HACH2212242 HACH2297255 705-0121 705-2032 705-0139 705-0141 HACH2293666 HACH2070042 HACH2070142 HACH2070242 705-0137 HACH2241732 HACH2241832 HACH2229726 HACH2257169 HACH2658300 HACH2257700 HACH2603100 HACH2603101 705-0116 HACH2352449 HACH2352512 HACH2368531 458-0045 458-0046 HACH2401066 HACH2401268 705-0152 705-2031 HACH2604299 705-0120 HACH2563000 HACH2459000 HACH2242300 705-2034 705-0560 HACH199526 705-0169 HACH2658448 HACH2293566 705-2038 HACH2790800 705-0199 HACH2076053 HACH2678500 HACH2678600 HACH2076049 HACH2754048 HACH2947553 HACHHPT310 HACH2368749 HACH2368964 HACH2368655 HACH2368855 HACH2105728 HACH2527025 HACH2658825
Reagents for DR800, DR900, DR2010, DR2500, DR2400, DR2800, DR3800, DR3900, DR5000, DR6000
Kits para análise ambiental
Reagents for photometric analysis either in field conditions or in the laboratory.
PP - Powder pillows - Simple and accurate addition of the reagents from a hermetically sealed aluminium foil pillow - ideal for measurement in difficult conditions.
AV - ACCUVAC - Sealed glass cuvettes containing a portioned amount of reagent. Simply immerse the tip of the accuvac in the sample, break it and the vacuum draws the sample into the cuvette.
IM - Immunoassay method of test.
RL- Rapid liquid test kit - Often used for trace level detection (e.g ultra low silica or total chlorine) and in conjunction with a flow cell when there is a high sample throughput.
RS - Reagent solution - Standard chemical reagents supplied in a bottle.
Atenção: * Suitable only for the DR 5000 UV-Vis Instrument.