Print…

Ácido clorídrico 36,5 - 38,0%, AR® ACS, Macron Fine Chemicals™

Fornecedor: Avantor Performance Materials
Perigo

Sinônimos: Hydrochloric acid solution, Muriatic acid

M.O.C. Category N

H613-27EA 0 EUR
H613-27
Ácido clorídrico 36,5 - 38,0%, AR® ACS, Macron Fine Chemicals™
Ácido clorídrico

ACS reagents meeting the requirements of the American Chemical Society Committee on Analytical Reagents. The specifications listed in this catalog are based on the ACS Reagent Chemicals Tenth Edition.  

Fórmula: HCl
Peso Molecular: 36.46 g/mol
Ponto de Fusão: –30 °C
Densidade: 1.18 g/cm³ (20 °C)
Temperatura de Armazenamento: Ambiente
Número MDL : MFCD00011324
Número CAS: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II

Encomendar

Especificações dos Resultados do Teste

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
ACS - Assay (HCl) 36.5 - 38.0 %
Appearance Passes Test
ACS - Color (APHA) ≤ 10
ACS - Ammonium (NH₄) ≤ 3 ppm
ACS - Arsenic (As) ≤ 0.005 ppm
ACS - Bromide (Br) ≤ 0.005 %
ACS - Extractable Organic Substances ≤ 5 ppm
ACS - Free Chlorine (as Cl) ≤ 1 ppm
ACS - Heavy Metals (as Pb) ≤ 0.1 ppm
ACS - Residue after Ignition ≤ 4 ppm
ACS - Sulfate (SO₄) ≤ 0.5 ppm
ACS - Sulfite (SO₃) ≤ 0.8 ppm
Trace Impurities - Aluminum (Al) ≤ 0.300 ppm
Trace Impurities - Boron (B) ≤ 0.100 ppm
Trace Impurities - Chromium (Cr) ≤ 0.200 ppm
Trace Impurities - Copper (Cu) ≤ 0.10 ppm
Trace Impurities - Gold (Au) ≤ 0.100 ppm
Trace Impurities - Iron (Fe) ≤ 0.1 ppm
Trace Impurities - Lead (Pb) ≤ 0.100 ppm
Trace Impurities - Magnesium (Mg) ≤ 0.300 ppm
Trace Impurities - Manganese (Mn) ≤ 0.300 ppm
Trace Impurities - Nickel (Ni) ≤ 0.1 ppm
Trace Impurities - Potassium (K) ≤ 0.300 ppm
Trace Impurities - Tin (Sn) ≤ 0.300 ppm
Trace Impurities - Titanium (Ti) ≤ 0.300 ppm
Trace Impurities - Zinc (Zn) ≤ 0.100 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR