Print…
Cole-Parmer® MP-250 Digital Melting Point Apparatus
Fornecedor: Cole-Parmer
03010-15 03010-41 03010-51 03010-59
ELET03010-15EA 2050 EUR
ELET03010-15 ELET03010-41 ELET03010-51 ELET03010-59
Cole-Parmer® MP-250 Digital Melting Point Apparatus
Aparelhos para determinação do ponto de fusão
Rotating viewing head ensures each user can analyse samples clearly.
  • View up to three samples simultaneously for greater efficiency
  • Height adjustable arm with viewing magnifier folds away for easy storage
  • Integral light and 8× magnifying lens enhance sample observation
  • Three audible beeps indicate when temperature is stable and ready for sample
  • 4×1 melt memory capacity
  • Adjustable object lens for sharp focus
The MP-250D series digital melting point apparatus allows you to easily view three capillary tube samples simultaneously through the 8× magnifyingMais informação do produto