Print…
Do Onions, Strawberries and Bananas Have DNA?
Fornecedor: EDVOTEK
S-75
EDVOS-75EA 75 EUR
EDVOS-75
Do Onions, Strawberries and Bananas Have DNA?
Materiais educationais Biology Educational Materials Genetics Learning Activities
Kit allows students to construct DNA models and then extract DNA from onions, strawberries or bananas.
  • For 10 lab groups
  • Complete in 30 minutes
Also required: 95 to 100% isopropyl alcohol, fruit and vegetables.
Also required: 95-100% isopropyl alcohol, fruit and vegetables.
Fornecimento: Kit includes instructions, DNA extraction buffer, DNA sample in capped test tube, transfer pipettes, pop beads, glass rod, DNA spooling rods, test tubes, and salt.